Anti-CD3, epsilon Hamster Monoclonal Antibody [clone: 5K3]
USBIC2254-11DX
- Antibody type:Primary
- Antigen symbol:CD3 epsilon
- Clonality:Monoclonal
- Clone:5K3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Hamster
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: CD3, epsilon
Clonality: Monoclonal
Clone: 5K3
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG Kappa
Reactivity: Mouse