Anti-ATF2 Rabbit Polyclonal Antibody
Catalog # USBIA0855-43W
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Activating transcription factor 211
- Antigen symbol:ATF211
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:ATF2
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
- Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry:1:50-1:100
Type: Primary
Antigen: ATF2 (activating transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat