Anti-SLC5A2 Rabbit Polyclonal Antibody
Catalog # USBI028121-30
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/glucose cotransporter), member 2
- Antigen symbol:Solute carrier family 5, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,5% BSA, 0,01% thimerosal, 30% glycerol.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5-4?g/ml
Type: Primary
Antigen: SLC5A2 (solute carrier family 5 (sodium/glucose cotransporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat