Anti-SLC13A3 Rabbit Polyclonal Antibody
Catalog # USBI138345
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3
- Antigen symbol:Solute carrier family 13, member 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 5 ?g/ml
Type: Primary
Antigen: SLC13A3 (solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat