Anti-SLC12A2 Rabbit Polyclonal Antibody
Catalog # USBI136419
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 12 (sodium/potassium/chloride transporter), member 2
- Antigen symbol:Solute carrier family 12, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 7. 5-10 ?g/ml
Type: Primary
Antigen: SLC12A2 (solute carrier family 12 (sodium/potassium/chloride transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human