Anti-SLC4A4 Mouse Monoclonal Antibody [clone: 1G2]
Catalog # USBI133498
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 4 (sodium bicarbonate cotransporter), member 4
- Antigen symbol:Solute carrier family 4, member 4
- Clonality:Monoclonal
- Clone:1G2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MSTENVEGKPSNLGERGRARSSTFLRVVQPMFNHSIFTSAVSPAAERIRFILGEEDDSPAPPQLFTELDELLAVDGQEMEWKETARWIKFEEKVEQGGERWSKPHVATLSLHSLFELRTCMEKGSIMLDREASSLPQLVEMIVDHQIETGLLKPELKDKVTYTLLRKHRHQTKKSNLRSLADIGKTVSSASRMFTNPDNGSPAMTHRNLTSSSLNDISDKPEKDQLKNKFMKKLPRDAEASNVLVGEVDFLDTPF
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SLC4A4 (solute carrier family 4 (sodium bicarbonate cotransporter), member 4)
Clonality: monoclonal
Clone: 1G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human