Anti-SLC13A3 Mouse Monoclonal Antibody [clone: 3A6]
USBI133401
- Antibody type:Primary
- Antigen name:solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3
- Antigen symbol:Solute carrier family 13, member 3
- Clonality:Monoclonal
- Clone:3A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:152 to 233
- Storage buffer:PBS, pH 7,2.
- Sequence:LPIANAILKSLFGQKEVRKDPSQESEENTAAVRRNGLHTVPTEMQFLASTEAKDHPGETEVPLDLPADSRKEDEYRRNIWK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: SLC13A3 (solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3)
Clonality: monoclonal
Clone: 3A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human