Anti-SGTA Rabbit Polyclonal Antibody
USBI133276
- Antibody type:Primary
- Antigen name:Small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha
- Antigen symbol:SGTA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 313
- Storage buffer:PBS, pH 7,2.
- Sequence:MDNKKRLAYAIIQFLHDQLRHGGLSSDAQESLEVAIQCLETAFGVTVEDSDLALPQTLPEIFEAAATGKEMPQDLRSPARTPPSEEDSAEAERLKTEGNEQMKVENFEAAVHFYGKAIELNPANAVYFCNRAAAYSKLGNYAGAVQDCERAICIDPAYSKAYGRMGLALSSLNKHVEAVAYYKKALELDPDNETYKSNLKIAELKLREAPSPTGGVGSFDIAGLLNNPGFMSMASNLMNNPQIQQLMSGMISGGNNPLGTPGTSPSQNDLASLIQAGQQFAQQMQQQNPELIEQLRSQIRSRTPSASNDDQQE
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: SGTA (small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse