Anti-SCNN1D Mouse Monoclonal Antibody [clone: 2F3]
USBI133032
- Antibody type:Primary
- Antigen name:sodium channel, non-voltage-gated 1, delta subunit
- Antigen symbol:SCNN1D
- Clonality:Monoclonal
- Clone:2F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:432 to 531
- Storage buffer:PBS, pH 7,2.
- Sequence:CFYRLYQDLETHRLPCTSRCPRPCRESAFKLSTGTSRWPSAKSAGWTLATLGEQGLPHQSHRQRSSLAKINIVYQELNYRSVEEAPVYSVPQLLSAMGS*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: SCNN1D (sodium channel, non-voltage-gated 1, delta subunit)
Clonality: monoclonal
Clone: 2F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human