Anti-SCN2B Mouse Monoclonal Antibody [clone: 2G11-C12]
Catalog # USBI133024
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:sodium channel, voltage-gated, type II, beta subunit
- Antigen symbol:SCN2B
- Clonality:Monoclonal
- Clone:2G11-C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MHRDAWLPRPAFSLTGLSLFFSLVPPGRSMEVTVPATLNVLNGSDARLPCTFNSCYTVNHKQFSLNWTYQECNNCSEEMFLQFRMKIINLKLERFQDRVEFSGNPSKYDVSVMLRNVQPEDEGIYNCYIMNPPDRHRGHGKIHLQVLMEEPPERDSTVAVIVGASVGGFLAVVILVLMVVKCVRRKKEQKLSTDDLKTEEEGKTDGEGNPDDGAK*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SCN2B (sodium channel, voltage-gated, type II, beta subunit)
Clonality: monoclonal
Clone: 2G11-C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human