Anti-SCN11A Mouse Monoclonal Antibody [clone: 6E1]
Catalog # USBI133023
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:sodium channel, voltage-gated, type XI, alpha subunit
- Antigen symbol:SCN11A
- Clonality:Monoclonal
- Clone:6E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1726 to 1792
- Storage buffer:PBS, pH 7,2.
- Sequence:TTTKRKEEERGAAIIQKAFRKYMMKVTKGDQGDQNDLENGPHSPLQTLCNGDLSSFGVAKGKVHCD*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SCN11A (sodium channel, voltage-gated, type XI, alpha subunit)
Clonality: monoclonal
Clone: 60
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human