Anti-ABCG1 Mouse Monoclonal Antibody [clone: 2E11]
Catalog # USBI122817
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family G, member 1
- Antigen symbol:ABCG1
- Clonality:Monoclonal
- Clone:2E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 lambda
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:AEMTEPKSVCVSVDEVVSSNMEATETDLLNGHLKKVDNNLTEAQRFSSLPRRAAVNIEFRDLSYSVPEGPWWRKKGYKTLLKGISGKFNSGELVAIMGPSGAGKSTLMNI
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ABCG1 (ATP-binding cassette, sub-family G (WHITE), member 1)
Clonality: monoclonal
Clone: 200000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Lambda
Reactivity: Human