Anti-SLC8A3 Rabbit Polyclonal Antibody
Catalog # USBI041990
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 8 (sodium/calcium exchanger), member 3
- Antigen symbol:Solute carrier family 8, member 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:592 to 622
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Type: Primary
Antigen: SLC8A3 (solute carrier family 8 (sodium/calcium exchanger), member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human