Anti-SLC4A7 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBI041958-PE
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 4, sodium bicarbonate cotransporter, member 7
- Antigen symbol:Solute carrier family 4, member 7
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:1193 to 1222
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: SLC4A7 (solute carrier family 4, sodium bicarbonate cotransporter, member 7)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human