Anti-SLC10A1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
USBI041782-APC
- Antibody type:Primary
- Antigen name:solute carrier family 10 (sodium/bile acid cotransporter), member 1
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Do not freeze!
Type: Primary
Antigen: SLC10A1 (solute carrier family 10 (sodium/bile acid cotransporter), member 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human