Anti-NCR3LG1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
USBI032367-FITC
- Antibody type:Primary
- Antigen name:Natural killer cell cytotoxicity receptor 3 ligand 1
- Antigen symbol:NCR3LG1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: Western Blot: 1:1000
Do not freeze!
Type: Primary
Antigen: NCR3LG1 (natural killer cell cytotoxicity receptor 3 ligand 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human