Order Entry
Finland
ContactUsLinkComponent
Anti-HLA-A Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 7G7F9]
Anti-HLA-A Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 7G7F9]
Catalog # USBI031022-HRP
Supplier:  US Biological
undefined
Anti-HLA-A Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 7G7F9]
Catalog # USBI031022-HRP
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    major histocompatibility complex, class I, A
  • Antigen symbol:
    HLA-A
  • Clonality:
    Monoclonal
  • Clone:
    7G7F9
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: HLA-A (major histocompatibility complex, class I, A)
Clonality: Monoclonal
Clone: 7G7F9
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse