Order Entry
Finland
ContactUsLinkComponent
Anti-CD44 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: Hermes-3]
Anti-CD44 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: Hermes-3]
Catalog # USBI030854-AP
Supplier:  US Biological
undefined
Anti-CD44 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: Hermes-3]
Catalog # USBI030854-AP
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD44 molecule (Indian blood group)
  • Antigen symbol:
    CD44
  • Clonality:
    Monoclonal
  • Clone:
    Hermes-3
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: Hermes-3
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human