Anti-SELL Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEL-14]
Catalog # ROCK200-502-N87
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Selectin L
- Antigen symbol:SELL
- Clonality:Monoclonal
- Clone:MEL-14
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:20343
- Antigen synonyms:Lyam-1|Lnhr|Lyam1|Ly-m22|Ly-22|LECAM-1|CD62L|AI528707|L-selectin
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
- Storage temperature:Cool
- Concentration:0,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Anti-CD62L Antibody (Monoclonal) was produced by repeated immunizations with CD62L antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Anti-CD62L is useful for immunochemistry, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: MEL-14
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse