Anti-SETDB2 Rabbit Polyclonal Antibody
ROCK200-401-N99S
- Antibody type:Primary
- Antigen name:SET domain, bifurcated 2
- Antigen symbol:SETDB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:0,98 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Setdb2 antibody was prepared from whole rabbit serum produced by repeated immunizations with a full legnth Protein corresponding to mouse Setdb2.
- Tested applications:Fluoresence ELISA
- Purification:Protein A purified
- Size:25 µl
- Pk:25 µl
Anti-Setdb2 antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 80.6 kDa in size corresponding to Setdb2 by western blotting.
Recommended Dilutions: ELISA: 1:2500 - 1:10000; Western Blot: 1:10000 - 1:100000
Type: Primary
Antigen: SETDB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human, Mouse