Order Entry
Finland
ContactUsLinkComponent
Anti-SETDB2 Rabbit Polyclonal Antibody
Anti-SETDB2 Rabbit Polyclonal Antibody
Catalog # ROCK200-401-N99
undefined
Anti-SETDB2 Rabbit Polyclonal Antibody
Catalog # ROCK200-401-N99

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SET domain, bifurcated 2
  • Antigen symbol:
    SETDB2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    0,98 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Setdb2 antibody was prepared from whole rabbit serum produced by repeated immunizations with a full legnth Protein corresponding to mouse Setdb2.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    Protein A purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Anti-Setdb2 antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 80.6 kDa in size corresponding to Setdb2 by western blotting.

Recommended Dilutions: ELISA: 1:2500 - 1:10000; Western Blot: 1:10000 - 1:100000

Type: Primary
Antigen: SETDB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human, Mouse