Order Entry
Finland
ContactUsLinkComponent
Anti-AKT1 Mouse Monoclonal Antibody (Biotin) [clone: 14E5.A2.B2.H9]
Anti-AKT1 Mouse Monoclonal Antibody (Biotin) [clone: 14E5.A2.B2.H9]
  ROCK200-306-I51
undefined
Anti-AKT1 Mouse Monoclonal Antibody (Biotin) [clone: 14E5.A2.B2.H9]
  ROCK200-306-I51

 

  • Antibody type:
    Primary
  • Antigen name:
    v-akt murine thymoma viral oncogene homolog 1
  • Antigen symbol:
    AKT1
  • Clonality:
    Monoclonal
  • Clone:
    14E5.A2.B2.H9
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Internal
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Gene ID:
    207
  • Antigen synonyms:
    CWS6|RAC|RAC-ALPHA|PRKBA|PKB|AKT|PKB-ALPHA
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,5 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-AKT1 Antibody was produced in mice by repeated immunizations with a synthetic peptide corresponding to internal residues of human AKT1 Protein followed by monoclonal development.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    Protein A purified
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Anti-AKT3 BIOTIN Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT3 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.A2.B2.H9
Conjugation: Biotin
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse, Rat, Human