Order Entry
Finland
ContactUsLinkComponent
Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 17H9.B4.H2]
Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 17H9.B4.H2]
Catalog # ROCK200-301-I41S
undefined
Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 17H9.B4.H2]
Catalog # ROCK200-301-I41S

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PROCALCITONIN
  • Clonality:
    Monoclonal
  • Clone:
    17H9.B4.H2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 lambda
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Procalcitonin Antibody was produced in mice prepared by repeated immunizations with full-length recombinant human Procalcitonin Protein.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    Protein A purified
  • Size:
    25 µl
  • Pk:
    25 µl

Specifications

About this item

Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 17H9.B4.H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 lambda
Reactivity: Human