Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 15F6.F6.F9]
ROCK200-301-I39
- Antibody type:Primary
- Antigen symbol:PROCALCITONIN
- Clonality:Monoclonal
- Clone:15F6.F6.F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 lambda
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Procalcitonin Antibody was produced in mice prepared by repeated immunizations with full-length recombinant human Procalcitonin Protein.
- Tested applications:Fluoresence ELISA
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml;
Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 15F6.F6.F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 lambda
Reactivity: Human