Order Entry
Finland
ContactUsLinkComponent
 
Anti-MMTV Rabbit Polyclonal Antibody
  ROCK100-401-P13S
undefined
Anti-MMTV Rabbit Polyclonal Antibody
  ROCK100-401-P13S

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MMTV
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Format:
    Antiserum
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    66mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    MMTV was prepared from whole rabbit serum produced by repeated immunizations with a full length sequence for mouse mammary tumor virus surface Protein tagged with His.
  • Tested applications:
    Fluoresence ELISA
  • Size:
    25 µl
  • Pk:
    25 µl

 

 

Anti-MMTV Antibody has been tested for use in western blotting. Specific conditions for reactivity should be optimized by the end user. Expect band approximately 41.2 kDa in size corresponding to MMTV surface Protein by western blotting.

Recommended Dilutions: ELISA: 1:10000-1:20000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: MMTV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Mouse