Anti-TET3 Rabbit Polyclonal Antibody
Catalog # ROCK100-401-N94S
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Tet Methylcytosine Dioxygenase 3
- Antigen symbol:TET3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:200424
- Antigen synonyms:hCG_40738
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:70 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogen:TET3 antibody was prepared from whole rabbit serum produced by repeated immunisations with a human TET-3 domain containing the N-terminal 160 amino acids of the Protein
- Tested applications:Fluoresence ELISA
- Purification:delipidation and defibrination
- Size:25 µl
- Pk:25 µl
Specifications
About this item
Tet3 antibody has been tested for use by ELISA and Western Blot. Specific conditions for reactivity should be optimised by the end user. Expect band at ~180 kDa.
Recommended Dilutions: ELISA: 1:50000; Western Blot: 1:5000.
Type: Primary
Antigen: TET3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human