Anti-IgG (H+L) Rabbit Polyclonal Antibody (Biotin)
ROCK614-4602
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Pig
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:2,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Swine IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Size:2 mg
- Pk:2 mg
This product has been assayed against 1.0 µg of Swine IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Pig