Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Texas Red®)
Catalog # ROCK613-4904
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG F(ab')2
- Clonality:Polyclonal
- Conjugation:Texas Red®
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Sheep
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:1,5 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Sheep IgG F(ab')2 fragment
- Purification:purified by immunoaffinity chromatography
- Size:1,5 mg
- Pk:1.500 µG
Specifications
About this item
Secondary Rabbit Anti-F(ab')2 Reacts with Sheep (Ovine)
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Sheep