Anti-CREB1 Rabbit Polyclonal Antibody
ROCK600-401-270
- Antibody type:Primary
- Antigen name:cAMP responsive element binding protein 1
- Antigen symbol:CREB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:1385
- Antigen synonyms:CREB
- Amino acid number:122 to 147
- Modification:phospho-Ser133
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:0,064 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:CREB phospho peptide corresponding to amino acid residues 122-147 of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Purification:affinity-purified
- Size:100 µl
- Pk:100 µG
Anti-CREB pS133 antibody reacts with phosphorylated human CREB at pS133 and shows minimal reactivity by western blot with non-phosphorylated CREB and minimal reactivity (1%) by ELISA against the non-phosphorylated form of the immunizing peptide.
Recommended Dilutions: ELISA: 1:1000 - 1:6000; Western Blot: 1:500 - 1:2000; Immunochemistry: 20 µg/ml;
Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human