Anti-APOB Goat Polyclonal Antibody
Catalog # ROCK600-101-111
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein B
- Antigen symbol:APOB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:338
- Antigen synonyms:apoB-100|FLDB|apoB-48|LDLCQ4
- Storage buffer:0,125 M Sodium Borate, 0,075 M Sodium Chloride, 0,005 M EDTA, pH 8.0, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,02 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:apoLipoProtein Type B was isolated from human plasma by density gradient centrifugation followed by HPLC purification.
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Anti-apoLipoProtein antibodies have been used for indirect trapping ELISA for quantitation of antigen in serum using a standard curve, for immunoprecipitation and for western blotting for highly sensitive qualitative analysis.
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:50 - 1:500; Immunoprecipitation: 1:100;
Type: Primary
Antigen: APOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human