Anti-EXOSC1 Mouse Polyclonal Antibody (Unconjugated) [clone: BM3]
ROCK200-301-277
- Antibody type:Primary
- Antigen name:Exosome Component 1
- Antigen symbol:EXOSC1
- Clonality:Monoclonal
- Clone:BM3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:51013
- Antigen synonyms:Csl4p|Ski4p|CGI-108|hCsl4p|SKI4|CSL4|p13
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:A BALB/c mouse was immunized with nuclei from Pokeweed mitogen stimulated human peripheral blood lymphocytes.
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
This antibody is suitable for flow cytometry and frozen section.
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: EXOSC1
Clonality: Polyclonal
Clone: BM3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse