Anti-ALPI Sheep Polyclonal Antibody
Catalog # ROCK100-6135
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:alkaline phosphatase, intestinal
- Antigen symbol:ALPI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- ImmunoPrecipitation:Yes
- Reactivity:Bovine
- Western blot:Yes
- Cross adsorption:none
- Format:Antiserum
- Form:Lyophilized
- Gene ID:280993
- Antigen synonyms:IAP
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2,
- Storage temperature:Cool
- Concentration:33,0 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Alkaline Phosphatase (Calf intestine)
- Purification:delipidation and defibrination
- Size:2 ml
- Pk:2 mL
Specifications
About this item
This product has been assayed against 1.0 µg of Alkaline Phosphatase (Calf intestine) in a standard ELISA using HRP conjugated Affinity Purified anti-Sheep IgG (H&L) (Rabbit) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Bovine