Order Entry
Finland
ContactUsLinkComponent
Anti-NOTCH2 Rabbit Polyclonal Antibody
Anti-NOTCH2 Rabbit Polyclonal Antibody
Catalog # ROCK100-401-408
undefined
Anti-NOTCH2 Rabbit Polyclonal Antibody
Catalog # ROCK100-401-408

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Notch 2
  • Antigen symbol:
    NOTCH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Format:
    Antiserum
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    4853
  • Antigen synonyms:
    AGS2|hN2|HJCYS
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
  • Storage temperature:
    Cool
  • Concentration:
    80 mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide corresponding to amino acid residues of human Notch 2 located near the N-terminal sequence of the cleaved N intracellular domain (NICD).
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Anti-Notch2 has a strong response was detected by ELISA against the immunizing peptide.This product was assayed against the peptide immunogen in a standard capture ELISA using Peroxidase conjugated anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:400 - 1:2000; Immunochemistry: 1:500;

Type: Primary
Antigen: NOTCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human