Anti-NFYA Rabbit Polyclonal Antibody
Catalog # ROCK100-401-100
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Nuclear transcription factor Y, alpha
- Antigen symbol:NFYA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:near the N-terminus
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:4800
- Antigen synonyms:CBF-A|CBF-B|NF-YA|HAP2
- Storage buffer:0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:90 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:NF-Y (A subunit specific) peptide corresponding to a region near the N-terminus of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Purification:delipidation and defibrination
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-NF-Y (A subunit) Antibody was assayed by immunoblot and found to be reactive against NF-Y (A subunit specific) at a dilution of 1:6,000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).
Recommended Dilutions: ELISA: 1:20000 - 1:100000; GelShift: Use 2.0 µl; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:400;
Type: Primary
Antigen: NFYA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human