Anti-PPI Goat Polyclonal Antibody
Catalog # ROCK100-1174
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Papain
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Reactivity:Carica papaya
- Western blot:Yes
- Cross adsorption:none
- Format:Antiserum
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:80 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Papain (Carica papaya)
- Purification:delipidation and defibrination
- Size:2 ml
- Pk:2 mL
Specifications
About this item
Primary Goat Anti-PAPAIN (Carica papaya) Reacts with Pan
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: PPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Carica papaya