Order Entry
Finland
ContactUsLinkComponent
Anti-G s alpha-Subunit, C-Terminal (385-394) Rabbit Polyclonal Antibody
Anti-G s alpha-Subunit, C-Terminal (385-394) Rabbit Polyclonal Antibody
Catalog # 371732-50
Anti-G s alpha-Subunit, C-Terminal (385-394) Rabbit Polyclonal Antibody
Catalog # 371732-50

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    G s alpha-Subunit
  • Antigen symbol:
    GSA subunit
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: G s alpha-Subunit, C-Terminal (385-394)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: