Anti-Trx-tag Mouse Monoclonal Antibody
Catalog # GENSA00180-40
Supplier: Genscript
Specifications
- Antibody type:Tag
- Antigen symbol:Trx-tag
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:40 µg
- Pk:40 µG
Specifications
About this item
Type: Tag
Antigen: Trx-tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: