Anti-CBS Rabbit Polyclonal Antibody
Catalog # BSBTPA1542
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cystathionine-beta-synthase
- Antigen symbol:CBS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:cystathionine beta-synthase|beta-thionase|serine sulfhydrase|HIP4|methylcysteine synthase
- Amino acid number:322 to 342
- Sequence:KWFKSNDEEAFTFARMLIAQE
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human CBS (322-342aa KWFKSNDEEAFTFARMLIAQE).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Cystathionine beta-synthase(CBS) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: CBS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human