Anti-Importin-8 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24751
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:importin 8
- Antigen symbol:IPO8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Gene ID:10526
- Antigen synonyms:RANBP8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Importin-8
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: