Anti-CHAF1A Mouse Monoclonal Antibody [clone: SS 1 1-13]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: CHAF1A (chromatin assembly factor 1, subunit A (p150))
Clonality: monoclonal
Clone: SS 1 1-13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB500-207SS
- NOVUNB500-207
- Antigen symbol:
- CHAF1A
- CHAF1A
- Antigen name:
- Chromatin assembly factor 1, subunit A (p150)
- Chromatin assembly factor 1, subunit A (p150)
- Antigen synonyms:
- CAF-1|CAF1P150|CAF1B|P150|CAF1
- CAF-1|CAF1P150|CAF1B|P150|CAF1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- SS 1 1-13
- SS 1 1-13
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 10036
- 10036
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice