Anti-Niemann-Pick type C1 Like-1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNB400-128H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Niemann-Pick type C1 like-1
- Antigen symbol:NPCL1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:29881
- Antigen synonyms:NPC11L1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Niemann-Pick type C1 Like-1
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat