Order Entry
Finland
ContactUsLinkComponent
Anti-SLC9A3 Mouse Monoclonal Antibody (DyLight® 650) [clone: 14D5]
Anti-SLC9A3 Mouse Monoclonal Antibody (DyLight® 650) [clone: 14D5]
Catalog # NOVUNB110-81529C
Supplier:  Novus Biologicals
Anti-SLC9A3 Mouse Monoclonal Antibody (DyLight® 650) [clone: 14D5]
Catalog # NOVUNB110-81529C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sodium/hydrogen exchanger 3
  • Antigen symbol:
    NHE3
  • Clonality:
    Monoclonal
  • Clone:
    14D5
  • Conjugation:
    DyLight® 650
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    6550
  • Antigen synonyms:
    sodium/hydrogen exchanger 3|NHE3|Na(+)/H(+) exchanger 3|NHE-3|solute carrier family 9 (sodium/hydrogen exchanger)|solute carrier family 9 member 3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: SLC9A3 (solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3)
Clonality: monoclonal
Clone: 14D5
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: