Order Entry
Finland
ContactUsLinkComponent
Anti-SLC9A3 Mouse Monoclonal Antibody [clone: 14D5]
Anti-SLC9A3 Mouse Monoclonal Antibody [clone: 14D5]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-SLC9A3 Mouse Monoclonal Antibody [clone: 14D5]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB110-81529
  • NOVUNB110-81529SS
  • Antigen symbol:
  • NHE3
  • NHE3
  • Antigen name:
  • Sodium/hydrogen exchanger 3
  • Sodium/hydrogen exchanger 3
  • Antigen synonyms:
  • sodium/hydrogen exchanger 3|NHE3|Na(+)/H(+) exchanger 3|NHE-3|solute carrier family 9 (sodium/hydrogen exchanger)|solute carrier family 9 member 3
  • sodium/hydrogen exchanger 3|NHE3|Na(+)/H(+) exchanger 3|NHE-3|solute carrier family 9 (sodium/hydrogen exchanger)|solute carrier family 9 member 3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 14D5
  • 14D5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 6550
  • 6550
  • Isotype:
  • IgG2a
  • IgG2a
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together