Anti-SLC9A3 Mouse Monoclonal Antibody [clone: 10A8]
Toimittaja: Novus Biologicals
Sertifikaatit
Tietoja tästä tuotteesta
Type: Primary
Antigen: SLC9A3 (solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3)
Clonality: monoclonal
Clone: 10A8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Tekniset tiedot
- Catalog No:
- NOVUNB110-74678SS
- NOVUNB110-74678
- Antigen symbol:
- NHE3
- NHE3
- Antigen name:
- Sodium/hydrogen exchanger 3
- Sodium/hydrogen exchanger 3
- Antigen synonyms:
- sodium/hydrogen exchanger 3|NHE3|Na(+)/H(+) exchanger 3|NHE-3|solute carrier family 9 (sodium/hydrogen exchanger)|solute carrier family 9 member 3
- sodium/hydrogen exchanger 3|NHE3|Na(+)/H(+) exchanger 3|NHE-3|solute carrier family 9 (sodium/hydrogen exchanger)|solute carrier family 9 member 3
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 10A8
- 10A8
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 6550
- 6550
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice