Anti-SLC9A8 Mouse Monoclonal Antibody (Biotin) [clone: 7A11]
Catalog # NOVUNB110-62091B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Sodium/hydrogen exchanger 8
- Antigen symbol:NHE8
- Clonality:Monoclonal
- Clone:7A11
- Conjugation:Biotin
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:23315
- Antigen synonyms:Sodium/hydrogen exchanger 8|Na (+)/H(+) exchanger 8|Solute carrier family 9 member 8|NHE-8|SLC9A8 antibody
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SLC9A8 (solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8)
Clonality: monoclonal
Clone: 7A11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat