Order Entry
Finland
ContactUsLinkComponent
Anti-SLC9A8 Mouse Monoclonal Antibody [clone: 7A11]
Anti-SLC9A8 Mouse Monoclonal Antibody [clone: 7A11]
Supplier:  Novus Biologicals
Certificates
Anti-SLC9A8 Mouse Monoclonal Antibody [clone: 7A11]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB110-62091SS
  • NOVUNB110-62091
  • Antigen symbol:
  • NHE8
  • NHE8
  • Antigen name:
  • Sodium/hydrogen exchanger 8
  • Sodium/hydrogen exchanger 8
  • Antigen synonyms:
  • Sodium/hydrogen exchanger 8|Na (+)/H(+) exchanger 8|Solute carrier family 9 member 8|NHE-8|SLC9A8 antibody
  • Sodium/hydrogen exchanger 8|Na (+)/H(+) exchanger 8|Solute carrier family 9 member 8|NHE-8|SLC9A8 antibody
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 7A11
  • 7A11
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 23315
  • 23315
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together