Anti-Dynamin Mouse Monoclonal Antibody [clone: 3G4B6]
Supplier: Novus Biologicals
Certificates
About this item
The Dynamin Antibody (3G4B6) from Novus Biologicals is a mouse monoclonal antibody to Dynamin. This antibody reacts with human, mouse. The Dynamin Antibody (3G4B6) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1: 200- 1: 1000
Type: Primary
Antigen: DNM
Clonality: Monoclonal
Clone: 3G4B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNB110-60491
- NOVUNB110-60491SS
- Antigen symbol:
- DNM
- DNM1
- Antigen name:
- Dynamin
- Dynamin 1
- Antigen synonyms:
- EIEE31|DNM
- EIEE31|DNM
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 3G4B6
- 3G4B6
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1759
- 1759
- Isotype:
- IgG2a
- IgG2a
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice