Order Entry
Finland
ContactUsLinkComponent
 
Anti-DNM1L Rabbit Polyclonal Antibody
 
undefined
Anti-DNM1L Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB110-55288
  • NOVUNB110-55288SS
  • Antigen symbol:
  • DRP1
  • DRP1
  • Antigen name:
  • Dihydropyrimidinase-related protein 1
  • Dihydropyrimidinase-related protein 1
  • Antigen synonyms:
  • dihydropyrimidinase-related protein 1|CRMP-1|DRP1|DPYSL1|unc-33-like phosphoprotein 3|dihydropyrimidinase-like 1|ULIP-3|dihydropyrimidinase related protein-1|DRP-1
  • dihydropyrimidinase-related protein 1|CRMP-1|DRP1|DPYSL1|unc-33-like phosphoprotein 3|dihydropyrimidinase-like 1|ULIP-3|dihydropyrimidinase related protein-1|DRP-1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 10059
  • 10059
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice