Order Entry
Finland
ContactUsLinkComponent
Anti-Eph Receptor A1 Rabbit Polyclonal Antibody
Anti-Eph Receptor A1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1573
Supplier:  Boster Bio
undefined
Anti-Eph Receptor A1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1573
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    EPH Receptor A1
  • Antigen symbol:
    EPHA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    EPHT|EPHA1|EPHT1|MGC163163|EPH
  • Amino acid number:
    963 to 976
  • UniProtKB:
    P21709
  • Sequence:
    HQKRILCSIQGFKD
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Eph receptor A1 (963-976aa HQKRILCSIQGFKD), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Ephrin type-A receptor 1(EPHA1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

EPHA1, EPH receptor A1, is a protein that in humans is encoded by the EPHA1 gene. This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. The EPHA1 gene is mapped to 7q34-q35. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene is expressed in some human cancer cell lines and has been implicated in carcinogenesis.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: Eph Receptor A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat