Order Entry
Finland
ContactUsLinkComponent
Anti-SR Rabbit Monoclonal Antibody
Anti-SR Rabbit Monoclonal Antibody
Catalog # ABCAAB140623-100
Supplier:  Abcam
Anti-SR Rabbit Monoclonal Antibody
Catalog # ABCAAB140623-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SR
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: